Learn More
IL-33, Mouse anti-Human, HRP, Clone: 6H496, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP230103H100UL
Catalog No. NB005485
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human Samples. It is validated for Western Blot.
Specifications
Specifications
IL-33
6H496
Western Blot
Purified
IL33
A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
Protein G purified
RUO
Primary
90865
Western Blot
HRP
PBS
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Mouse
IgG1 Kappa
100 uL
Store at 4°C in the dark.
Monoclonal
Human