Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33 Mouse anti-Human, Janelia Fluor™ 646, Clone: 6H617, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227333JF646

Catalog No. N227333J646


Add to cart

Description

Description

IL-33 Monoclonal antibody specifically detects IL-33 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

IL-33
6H617
Western Blot, Flow Cytometry
Conjugated, Purified
IL33
Full length recombinant protein
Protein G purified
Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary
90865.0
Flow Cytometry, Western Blot
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit