Learn More
IL-33 Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: 1C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP231145AF594
Catalog No. NP23114A94
Add to cart
Description
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
IL-33
1C5
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
IL33
Synthetic peptide corresponding to an internal sequence of the human IL33 protein (between amino acids 100-270) [UniProtNo. O95760]
0.1 mL
Primary
90865