Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Monoclonal Antibody (578416)
IL-34 Monoclonal Antibody (578416)
Supplier: Thermo Scientific MA524259
Description

Specifications
IL-34 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6ZMJ4 | |
Il34 | |
Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Neutralization | |
578416 | |
Unconjugated | |
Il34 | |
2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
Mouse | |
Protein A/G | |
RUO | |
146433 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction