Learn More
IL-34 Mouse anti-Human, Alexa Fluor™ 405, Clone: 36W18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221982AF405
Catalog No. NP221982A40
Add to cart
Description
Description
IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for ELISA.
Specifications
Specifications
IL-34
36W18
ELISA
C16orf77, IL-34, interleukin 34
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
IL34
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant IL-34. Mouse anti human IL-34 Q6ZMJ4.
0.1 mL
Primary
146433