Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-34 Mouse anti-Human, Alexa Fluor™ 750, Clone: 36W18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221982AF750

Catalog No. NP221982A70


Add to cart

Description

Description

IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for ELISA.
Specifications

Specifications

IL-34
36W18
ELISA
Conjugated, Purified
IL34
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant IL-34. Mouse anti human IL-34 Q6ZMJ4.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
C16orf77, IL-34, interleukin 34
Mouse
IgG1
0.1 mL
Primary
146433.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit