Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-36 alpha/IL-1F6 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
$184.44 - $509.24
Specifications
Antigen | IL-36 alpha/IL-1F6 |
---|---|
Clone | 1087L |
Dilution | Western Blot 1 ug/mL, Neutralization 0.2-1.2 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB10781SP
|
R&D Systems
MAB10781SP |
Unconjugated | 25 μg |
Each for $184.44
|
|
||||
MAB10781100
|
R&D Systems
MAB10781100 |
Unconjugated | 100 μg |
Each for $509.24
|
|
||||
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for Western Blot, Neutralization.Specifications
IL-36 alpha/IL-1F6 | |
Western Blot 1 ug/mL, Neutralization 0.2-1.2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
E. coli-derived recombinant human IL-36 alpha/IL-1F6 Lys6-Phe158 Accession # Q9UHA7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. |
1087L | |
Western Blot, Neutralization | |
Unconjugated | |
Rabbit | |
Human | |
Q9UHA7 | |
27179 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title