Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 beta/IL-1F8 Rabbit anti-Mouse, DyLight 405, Clone: 110, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290786V

Catalog No. NB041038


Add to cart

Description

Description

IL-36 beta/IL-1F8 Monoclonal antibody specifically detects IL-36 beta/IL-1F8 in Mouse samples. It is validated for Western Blot, ELISA
Specifications

Specifications

IL-36 beta/IL-1F8
110
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Biologically Active Proteins
Primary
27177
ELISA, Western Blot
DyLight 405
50mM Sodium Borate
family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Recombinant Mouse IL-36 beta/IL-1F8 Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit