Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 gamma/IL-1F9 Rabbit anti-Human, Clone: 13, Novus Biologicals™

Rabbit Monoclonal Antibody

$321.00 - $493.00

Specifications

Antigen IL-36 gamma/IL-1F9
Clone 13
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:10000-1:20000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020046
View Documents
Novus Biologicals
NBP289281100UL
100 μg
Each for $493.00
Add to cart
 
NB020047
View Documents
Novus Biologicals
NBP28928150UL
50 μg
Each for $321.00
Add to cart
 
Description

Description

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

IL-36 gamma/IL-1F9
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10000-1:20000
Monoclonal
Purified
RUO
Human
IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Recombinant Human IL-36 gamma/IL-1F9 Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
13
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Biologically Active Proteins
0.2 um filtered solution in PBS
56300
IgG
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit