Learn More
IL-37/IL-1F7 Mouse anti-Human, Alexa Fluor 647, Clone: MM0386-4C33, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211686AF647
Catalog No. NB211686647
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IL-37/IL-1F7 Monoclonal antibody specifically detects IL-37/IL-1F7 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-37/IL-1F7
MM0386-4C33
Western Blot
Purified
IL37
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-1F7.
Protein G purified
Cytokine Research
Primary
27178
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
FIL1, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human