Learn More
IL-38/IL-1F10 Rat anti-Mouse, Alexa Fluor 700, Clone: 798036, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB7774N100UG
Catalog No. FAB7774N100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-38/IL-1F10 Monoclonal antibody specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
IL-38/IL-1F10
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q8R459
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Rat
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse IL-38/IL-1F10 in ELISA and Western Blot.
Flow Cytometry
798036
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Purified
IL1F10
E. coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession # Q8R459
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84639
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)