Learn More
Medchemexpress LLC Interleukin-3 receptor subunit alpha (IL-3R alpha/CD123) | >95.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70074750UG
Encompass_Preferred
Recombinant human interleukin-3 receptor subunit alpha (IL-3R alpha/CD123) is a glycosylated, biotinylated protein produced in HEK293 cells with C-terminal His and Avi tags. It corresponds to residues T19-R305 (accession P26951-1) and is provided for receptor-binding studies, immunoassays, and biochemical characterization.
  • Recombinant human protein suitable for biochemical and cell-based assays.
  • Avi-tag biotinylated for streptavidin-based detection or capture.
  • C-terminal His tag facilitates purification and detection.
  • High purity (>95%) as assessed by reducing SDS-PAGE.
  • Glycosylated; molecular weight approximately 55-65 kDa.
  • Suitable for ELISA, binding assays, and antibody characterization.
  • Available in small pack sizes including 50 μg for assay workflows.
Catalog No. 50-001-77256