Learn More
IL-3R alpha/CD123 Mouse anti-Human, Alexa Fluor™ 750, Clone: 18N6B12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227225AF750
Catalog No. NP22722A70
Add to cart
Description
Description
IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
IL-3R alpha/CD123
18N6B12
Western Blot, Flow Cytometry
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
IL3RA
A portion of amino acids 250-300 of human CD123 was used as the immunogen for this antibody.
0.1 mL
Primary
3563