Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R alpha/CD123 Rabbit anti-Human, Clone: 17, Novus Biologicals™
Rabbit Monoclonal Antibody
$340.00 - $521.00
Specifications
Antigen | IL-3R alpha/CD123 |
---|---|
Clone | 17 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
IL-3R alpha/CD123 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
Recombinant Human IL-3R alpha/CD123 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
17 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
0.2 um filtered solution in PBS | |
3563 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title