Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R beta Antibody (130705), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5492X
Description
IL-3R beta Monoclonal antibody specifically detects IL-3R beta in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-3R beta | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1439 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
130705 | |
Flow Cytometry, CyTOF-ready | |
AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3RB, Il3rb2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta , His23-Trp440, Accession # P26954 | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction