Learn More
IL-4 Rat anti-Human, BV421, Clone: MP4-25D2, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566274
Catalog No. BDB566274
Add to cart
Description
Description
IL-4 Monoclonal antibody specifically detects IL-4 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
IL-4
MP4-25D2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Interleukin-4; IL4; BCGF-1; BSF-1; Lymphocyte stimulatory factor 1
Purified Recombinant Human IL-4
25 Tests
Cell Biology
Primary
Human
Flow Cytometry, Flow Cytometry (Intracellular Staining)
Brilliant Violet 421
Purified
Rat
Rat IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.