Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4I1 Antibody (CL9071), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP288929
Description
IL-4I1 Monoclonal specifically detects IL-4I1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-4I1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
259307 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CL9071 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
FIG1, Fig-1 protein, hFIG1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: SFRRPFWREEHIEGGHSNTDRPSR | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction