Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4R alpha Rabbit anti-Human, Alexa Fluor 350, Clone: 209, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289472AF350

Catalog No. NB066993


Add to cart

Description

Description

IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

IL-4R alpha
209
Flow Cytometry
Purified
Rabbit
IgG
0.1 mL
Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell
Primary
3566
Flow Cytometry
Alexa Fluor 350
50mM Sodium Borate
CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Recombinant Human IL-4R alpha Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit