Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Rat anti-Human, Mouse, Clone: TRFK5R, R&D Systems™

Recombinant Monoclonal Antibody

$148.35 - $369.00

Specifications

Antigen IL-5
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Format Purified
Clone TRFK5R
Applications ELISA, Neutralization
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB405R100 View Documents R&D Systems
MAB405R100
100 ug Each for $369.00
Add to cart
 
MAB405RSP View Documents R&D Systems
MAB405RSP
25 ug Each for $148.35
Add to cart
 
Description

Description

IL-5 is a disulfide-linked homodimeric cytokine that is secreted by T cells. IL-5 promotes the proliferation, differentation, and activation of eosinophils. It binds to a receptor complex consisting of one IL-5 specific α chain and one non-binding common β chain that is shared with the receptors for GM-CSF and IL-3.
Specifications

Specifications

IL-5
Purified
ELISA, Neutralization
Rat
Protein A or G purified from cell culture supernatant
RUO
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
TRFK5R
Unconjugated
IgG1
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit