Learn More
IL-6 Monoclonal Antibody (MQ2-13A5), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$358.65
Specifications
Antigen | IL-6 |
---|---|
Clone | MQ2-13A5 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-169-28
|
Invitrogen
56706942 |
100 Tests |
Each of 1
for $358.65
Save 4% Reg : |
|
|||||
Description
Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28 kDa cytokine secreted by a variety of cell types. Applications Reported: This MQ2-13A5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MQ2-13A5 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry.Specifications
IL-6 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P05231 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MQ2-13A5 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Rat | |
Human | |
3569 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.