Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Rat anti-Human, Monkey, Rhesus Macaque, Alexa Fluor™ 350, Clone: MQ2-13A5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233047AF350

Catalog No. NP233047A30


Add to cart

Description

Description

IL-6 Monoclonal antibody specifically detects IL-6 in Human, Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

IL-6
MQ2-13A5
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
IL6
COS-expressed human IL-6
Protein A or G purified
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Primary
3569.0
Human, Monkey, Rhesus Macaque
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Rat
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human/Rhesus/Cynomolgus IL-6
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit