Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Rat anti-Human, Monkey, Rhesus Macaque, PE-Cyanine7, Clone: MQ2-13A5, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP233047PECY7
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human, Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry.Specifications
IL-6 | |
MQ2-13A5 | |
Flow Cytometry | |
Conjugated, Purified | |
IL6 | |
COS-expressed human IL-6 | |
Protein A or G purified | |
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
Primary | |
3569.0 | |
Human, Monkey, Rhesus Macaque |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
Rat | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human/Rhesus/Cynomolgus IL-6 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title