Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6R alpha Antibody (Hu137), mFluor Violet 450 SE, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB10346MFV450
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
| IL-6R alpha | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Human | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, Neutralization, CyTOF | |
| Hu137 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Human IL-6R alpha | |
| RUO | |
| 3570 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction