Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7 Mouse anti-Human, Clone: AH31-34, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP267278
Description
IL-7 Monoclonal specifically detects IL-7 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IL-7 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
IL-7interleukin-7, interleukin 7 | |
Mouse | |
Protein A purified | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
3574 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
AH31-34 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.2% Sodium Azide | |
IL7 | |
Full length protein | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction