Learn More
IL-7R alpha/CD127 Rat anti-Mouse, Alexa Fluor™ 532, Clone: 73N4E10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225263AF532
Catalog No. NP22263A32
Add to cart
Description
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
IL-7R alpha/CD127
73N4E10
Western Blot
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Rat
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
IL7R
Amino acids 145-235 of mouse IL-7R/ CD127 were used as the immunogen for the antibody.
0.1 mL
Primary
3575