Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-7R alpha/CD127 Rat anti-Mouse, Alexa Fluor™ 594, Clone: 73N4E10, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225263AF594

Catalog No. NP22263A94


Add to cart

Description

Description

IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IL-7R alpha/CD127
73N4E10
Western Blot
Conjugated, Purified
IL7R
Amino acids 145-235 of mouse IL-7R/ CD127 were used as the immunogen for the antibody.
Protein G purified
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
Primary
3575.0
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Rat
IgG Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit