Learn More
IL-7R alpha/CD127 Mouse anti-Rat, Alexa Fluor 405, Clone: 717519, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8484V100UG
Catalog No. FAB8484V100
Add to cart
This item is not returnable. View return policy
Description
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-7R alpha/CD127
717519
Flow Cytometry 0.25-1μg/106 cells
NP_001099888
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession # NP_001099888
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects rat IL-7 R alpha/CD127in direct ELISAs.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Mouse
IgG2a
100μg
Primary
3575
Rat