Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (AH9R7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236513B
Description
IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for ELISA.Specifications
IL-9R | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
IL9R | |
Human CD129-transfected cell line | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody AH9R7 recognizes CD129 IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons. | |
Store at 4C in the dark. | |
IgG2b κ |
ELISA | |
AH9R7 | |
ELISA | |
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
Mouse | |
Protein A purified | |
RUO | |
3581 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction