Learn More
IL-9R Mouse anti-Human, DyLight 680, Clone: AH9R7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236513FR
Catalog No. NBP236513FR
Add to cart
Description
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
Specifications
IL-9R
AH9R7
Flow Cytometry, ELISA
Purified
IL9R
Human CD129-transfected cell line
Protein A purified
RUO
Primary
3581
Human
ELISA, Flow Cytometry
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Mouse
IgG2b Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody AH9R7 recognizes CD129 IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons.