Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9R Mouse anti-Human, PerCP, Clone: AH9R7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236513PCP

Catalog No. NBP236513JP


Add to cart

Description

Description

IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

IL-9R
AH9R7
Flow Cytometry, ELISA
Purified
IL9R
Human CD129-transfected cell line
Protein A purified
RUO
Primary
3581
Human
ELISA, Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Mouse
IgG2b Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody AH9R7 recognizes CD129 IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit