Learn More
IL-9R Rat anti-Mouse, DyLight 488, Clone: RM0115-12F43, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122493G
Catalog No. NBP122493G
Add to cart
Description
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
IL-9R
RM0115-12F43
Western Blot
Purified
IL9R
Mouse recombinant protein of the extracellular domain of IL-9 R
Protein G purified
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-9 R.
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Rat
IgG2
0.1 ml
Primary
3581
Mouse