Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Mouse anti-Human, Alexa Fluor™ 350, Clone: AH9R7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236513AF350
Description
IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
IL-9R | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
IL9R | |
Human CD129-transfected cell line | |
0.1 mL | |
3581.0 | |
Human | |
IgG2b κ |
Flow Cytometry, ELISA | |
AH9R7 | |
Flow Cytometry, ELISA | |
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
Mouse | |
Protein A purified | |
Primary | |
The mouse monoclonal antibody AH9R7 recognizes CD129/ IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons. | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title