Learn More
IL10RB Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 193871AP
Catalog No. 50-173-2393
Add to cart
Description
Description
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Specifications
Specifications
IL10RB
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q08334
CDW210B, CRF2 4, CRFB4, D21S58, D21S66, IL 10 receptor subunit beta, IL 10R subunit 2, IL 10R subunit beta, IL 10R2, IL 10RB, IL10RB, IL-10Rbeta, interleukin 10 receptor, beta
Rabbit
IgG
150 μL
-20°C
Primary
3588
Western Blot
Unconjugated
IL10RB
Liquid
IL10RB
IL10RB Fusion Protein Ag7468
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human