Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ IL12 (p70) (Human) ELISA Kit
Sandwich enzyme immunoassay for quantitative measurement of humanIL12 (p70).
Manufacturer:  Abnova™ KA4807
Catalog No. 89-948-614
Description
Description
  • Used for the in vitro quantitative determination of human IL-12 p70 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-12 p70
Specifications
Specifications
Colorimetric
7.8125 to 500 pg/mL
IL12 (p70) (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL12 (p70).
3592
ELISA Kit
100 μL
Human
P29459;P29460
Biotin
Quantification
Human IL-12 p70 microtiter plate
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Culture Supernatant, Plasma, Serum