Learn More
Aviva Systems Biology Corp IL15 Antibody - N-terminal region (ARP55616_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55616_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL15. Alternate names: IL-15, MGC9721ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-61554