Learn More
IL15RA Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 167441AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-1154
Add to cart
Description
Description
Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
Specifications
Specifications
IL15RA
Polyclonal
Unconjugated
IL15RA
IL 15 receptor subunit alpha, IL 15R alpha, IL 15RA, IL15RA, interleukin 15 receptor, alpha, sIL 15 receptor subunit alpha, sIL 15R alpha, sIL 15RA
Rabbit
Antigen Affinity Chromatography
RUO
3601
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13261
IL15RA
IL15RA Fusion Protein Ag10151
150 μL
Primary
Human
Antibody
IgG