Learn More
IL17B Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P248BT1MG

Description
AA Sequence of recombinant protein: MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17B. Anti-Human IL-17B-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-17B by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-17B (500-P248) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-17B. Western Blot: To detect hIL-17B by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-17B is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL17B Polyclonal antibody specifically detects IL17B in Human samples. It is validated for ELISA, Western BlotSpecifications
IL17B | |
Polyclonal | |
PBS with no preservative | |
Q9UHF5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27190 | |
-20°C |
ELISA, Western Blot | |
Biotin | |
IL17B | |
1110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7 | |
E.coli-derived Recombinant Human IL-17B | |
1 mg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.