Sino Biological™ IL17RC Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain of human IL17RC isoform 3 (Met 1- Ala 454) fused to Fc region of human IgG1 at C-terminus

Manufacturer: Sino Biological™ 11747H02H250

 View more versions of this product

Catalog No. 11-747-H02H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Protein Family Cytokines and Receptors
    Format Lyophilized
    Molecular Weight 75.3kd
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 50μg
    Source Human
    Structural Form Truncated, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Interleukin 17 receptor C
    Gene Symbol IL17RC
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Expression System Mammalian
    Product Type L17RC Recombinant Human Protein

    Expressed from DNA sequence encoding extracellular domain of human IL17RC isoform 3 (Met 1- Ala 454) fused to Fc region of human IgG1 at C-terminus

    • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit