Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL17RD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586992
Description
Purity is > 95% by SDS-PAGE.
Feedback inhibitor of fibroblast growth factor mediated Ras-MAPK signaling and ERK activation. May inhibit FGF-induced FGFR1 tyrosine phosphorylation. Regulates the nuclear ERK signaling pathway by spatially blocking nuclear translocation of activated ERK without inhibiting cytoplasmic phosphorylation of ERK. Mediates JNK activation and may be involved in apoptosis. Might have a role in the early stages of fate specification of GnRH-secreting neurons.
Specifications
IL17RD | |
Polyclonal | |
Unconjugated | |
Il17rd | |
2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171463, 498576, 54756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8JZL1, Q8NFM7 | |
Il17rd | |
Recombinant fusion protein of human IL17RD. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction