Learn More
IL18BP Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Supplier: Pepro Tech Inc 500P153GBT1MG

Description
Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine IL-18BP. Anti-Murine IL-18BP-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine IL-18BP by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-18BP (500-P153G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-18BP. Western Blot: To detect mIL-18BP by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-18BP is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL18BP Polyclonal antibody specifically detects IL18BP in Mouse samples. It is validated for ELISA, Western BlotSpecifications
IL18BP | |
Polyclonal | |
PBS with no preservative | |
Q9Z0M9 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16068 | |
-20°C |
ELISA, Western Blot | |
Biotin | |
IL18BP | |
Igifbp; Il18bp; IL-18BP; IL18BPa; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18 binding protein; interleukin-18-binding protein; MC51L-53L-54L homolog gene product; MC54L; Tadekinig-alfa | |
E.coli-derived Recombinant Murine IL-18BP | |
1 mg | |
Primary | |
Mouse | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.