Learn More
Novus Biologicals™ IL18R1 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2795215PLATES
 View more versions of this product
Catalog No. NB010082
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse IL18R1.
Specifications
Specifications
Sandwich ELISA
HRP
6.25 pg/mL
8809
RUO
IL18R1
5 Plates
6.25-400 pg/mL
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Antibody Pairs
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse IL18R1.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-IL18R1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse IL18R1 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 40 ng of recombinant Mouse IL18R1.
  • Storage is content dependent.
    Rabbit
    Mouse
    IL18R1
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.