Learn More
Invitrogen™ IL18R1 Monoclonal Antibody (70625.111)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA124813
Description
In Neutralization assays at a concentration of 0.12 μg/mL, this antibody will neutralize 50% of the bioactivity due to 50 ng/mL recombinant human IL18 using activated human T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
IL18R1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q13478 | |
Il18r1 | |
Recombinant, full length human IL18R1 protein expressed in mouse NSO cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
70625.111 | |
Unconjugated | |
Il18r1 | |
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
Mouse | |
purified | |
RUO | |
8809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.