Learn More
IL18R1 Mouse anti-Human, DyLight 550, Clone: MM0380-8H22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211679R
Catalog No. NBP211679R
Add to cart
Description
Description
IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IL18R1
MM0380-8H22
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IL18R1
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-18 Ra extracellular domain
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Mouse
IgG1
0.1 ml
Primary
8809