Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Rabbit anti-Cynomolgus Monkey, FITC, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298456F
Description
IL18R1 Polyclonal specifically detects IL18R1 in Cynomolgus Monkey samples. It is validated for ELISA.Specifications
IL18R1 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
8809 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
Produced in rabbits immunized with purified, recombinant IL18R1 (F7H8F9; Met1-Lys315). | |
0.1 mL | |
Primary | |
Cynomolgus Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title