Learn More
ACROBiosystems Recombinant Protein;Human IL-1 beta / IL-1F2 Protein, Tag Free, low endotoxin;E.coli;50UG;ILB,IL1B,IL-1B,IL-1,IL-1BETA,IL1F2,IL-1β,IL1β,IL1,Interleukin-1 beta,Interleukin-1beta,Interleukin1beta,IL1BETA,IL1 BETA,Interleukin-1,Interleukin
Supplier:  ACROBiosystems ILBH411050UG
Encompass_Preferred
Recombinant Protein;Human IL-1 beta / IL-1F2 Protein, Tag Free, low endotoxin;50UG;Human IL-1 beta, Tag Free (ILB-H4110) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # NP_000567).;This protein carries no "tag". The protein has a calculated MW of 17.5 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-1 beta, Tag Free (Cat. No. ILB-H4110) at 10μg/mL (100 μL/well) can bind Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) with a linear range of 0.039-0.15 μg/mL (QC tested).
Catalog No. 50-165-7641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.