Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1F6 Mouse anti-Human, Clone: OTI3A12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501427S

Catalog No. 50-167-2570


Add to cart

Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

IL1F6
Monoclonal
0.42 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UHA7
FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6
Mouse
IgG1
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27179
Immunocytochemistry, Immunofluorescence, Western Blot
OTI3A12
Unconjugated
IL36A
Liquid
IL36A
Full length human recombit protein of human IL1F6 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit