Learn More
IL1F9 Rat anti-Human, Alexa Fluor 647, Clone: MM0388-2P38, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211688AF647
Catalog No. NB211688647
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IL-36 gamma/IL-1F9 Monoclonal antibody specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL1F9
MM0388-2P38
Western Blot
Purified
IL36G
Produced from a hybridoma (mouse myeloma fused with spleen cells from a Rat immunized with human recombinant protein of IL-1F9.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
IL1E, IL-1F9IL-1 epsilon, IL1H1IL-1-epsilon, IL-1H1IL-1-related protein 2, IL1RP2IL-1 related protein 2, IL-1RP2IL-1(EPSILON), interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1
Rat
IgG2
0.1 ml
Primary
56300