Sino Biological™ IL1R1 Recombinant Rat Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding rat IL1R1 extracellular domain (Met 1-Lys 352) fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 80028R08H50

 View more versions of this product

Catalog No. 80-028-R08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 38.5kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Rat
    Structural Form Truncated, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Interleukin 1 Receptor Type 1
    Gene Symbol IL1R1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Cytokines and Receptors, CD Proteins
    Expression System Mammalian
    Product Type IL1R1 Recombinant Rat Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 25663

    Expressed from DNA sequence encoding rat IL1R1 extracellular domain (Met 1-Lys 352) fused to polyhistidine tag at C-terminus

    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit