Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1RAPL1 Antibody (MM0353-3R16), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211648AF700
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot.Specifications
IL1RAPL1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL1RAPL1 | |
Human recombinant IL1RAPL1 (extracellular domain) | |
0.1 mL | |
11141 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
MM0353-3R16 | |
Western Blot | |
IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction