Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1RAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 216091AP150UL
Description
The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signalling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.Specifications
IL1RAPL1 | |
Polyclonal | |
Unconjugated | |
IL1RAPL1 | |
IL 1 RAPL 1, IL 1RAPL 1, IL1R8, IL1RAPL, IL1RAPL 1, IL1RAPL1, MRX10, MRX21, MRX34, Oligophrenin 4, OPHN4, TIGIRR 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11141, 317553, 331461 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59823, P59824, Q9NZN1 | |
Il1rapl1 | |
IL1RAPL1 Fusion Protein Ag16252 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction